Learn More
AdipoGen Bromopropyl-N-benzoxadiazol-am
SDP
Supplier:  AdipoGen CDXB0268M500
Encompass_Preferred
Chemical. CAS 1246965-49-0. Formula C9H9BrN4O3. MW 301.1. Synthetic. Building block for synthesis. Used for the preparation of fluorescent probes.
Catalog No. 50-256-1614