Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromopyrogallol Red, TCI America™
SDP

Catalog No. D02075G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D02075G 5 g
D02071G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D02075G Supplier TCI America Supplier No. D02075G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 16574-43-9
Molecular Formula C19H10Br2O8S
Molecular Weight (g/mol) 558.15
MDL Number MFCD00148865
InChI Key QFXYYBXMARTXHI-UHFFFAOYSA-N
Synonym Dibromopyrogallolsulfonphthaleine
PubChem CID 85495
IUPAC Name 2',7'-dibromo-3',4',5',6'-tetrahydroxyspiro[2,1λ⁶-benzoxathiole-3,9'-xanthene]-1,1-dione
SMILES OC1=C(O)C2=C(C=C1Br)C1(OS(=O)(=O)C3=CC=CC=C13)C1=CC(Br)=C(O)C(O)=C1O2

Specifications

Quantity 5 g
Physical Form Crystal/Powder
Formula Weight 558.15
Chemical Name or Material Bromopyrogallol Red
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.