Learn More
Selleck Chemical LLC Bromopyruvic acid-25mg
SDP
Catalog No. 502389925
Encompass_Preferred
Bromopyruvic acid (3-Bromopyruvic acid, 3-Bromopyruvate, Bromopyruvic acid, Hexokinase II Inhibitor II, 3-BP) is a hexokinase II inhibitor with Ki of 2.4 mM for glycolysis/hexokinase inhibition. It is inhibitor of tumour cell energy metabolism and chemopotentiator of platinum drugs.
Catalog No. 50-238-9925 Supplier Selleck Chemical LLC Supplier No. S542625MG
Bromopyruvic acid (3-Bromopyruvic acid, 3-Bromopyruvate, Bromopyruvic acid, Hexokinase II Inhibitor II, 3-BP) is a hexokinase II inhibitor with Ki of 2.4 mM for glycolysis/hexokinase inhibition. It is inhibitor of tumour cell energy metabolism and chemopotentiator of platinum drugs.