Learn More
Selleck Chemical LLC Bromopyruvic acid (3-BP) S5426-1g
SDP
Catalog No. 503110251
Encompass_Preferred
Bromopyruvic acid (3-BP) is a hexokinase II inhibitor with Ki of 2 4 mM for glycolysis/hexokinase inhibition It is inhibitor of tumour cell energy metabolism and chemopotentiator of platinum drugs
Catalog No. 50-311-0251 Supplier Selleck Chemical LLC Supplier No. S54261G
Bromopyruvic acid (3-BP) is a hexokinase II inhibitor with Ki of 2 4 mM for glycolysis/hexokinase inhibition It is inhibitor of tumour cell energy metabolism and chemopotentiator of platinum drugs