Learn More
Cayman Chemical Bromosporine, Purity: ≥98%, CAS: 1619994-69-2, 10 mg
SDP
Catalog No. 502954653
Encompass_Preferred
A non-selective bromodomain inhibitor that at 1 µM has been shown to accelerate FRAP recovery of BRD4 and CREBBP bromodomains in HeLa cells
Catalog No. 50-295-4653 Supplier Cayman Chemical Supplier No. 1411910 MG
A non-selective bromodomain inhibitor that at 1 µM has been shown to accelerate FRAP recovery of BRD4 and CREBBP bromodomains in HeLa cells