Learn More
Eurofins Discoverx Bromosporine, 10 mg. (92-1239)
Catalog No. 501999760
Encompass
Bromosporine is a synthetic small molecule pan-bromodomain inhibitor developed by the Structural Genomics Consortium for research. It is shown to accelerate FRAP recovery of BRD4 and CREBBP in cells at 1uM concentration. Bromodomains are implicated in multiple cellular processes, including inflammatory gene expression, mitosis, and viral/host interaction. Provided as 10 mg of dry powder. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9760 Supplier Eurofins Discoverx Supplier No. 921239
Bromosporine is a synthetic small molecule pan-bromodomain inhibitor developed by the Structural Genomics Consortium for research. It is shown to accelerate FRAP recovery of BRD4 and CREBBP in cells at 1uM concentration. Bromodomains are implicated in multiple cellular processes, including inflammatory gene expression, mitosis, and viral/host interaction. Provided as 10 mg of dry powder. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.