Learn More
Selleck Chemical LLC Bromosporine 5mg
SDP
Supplier:  Selleck Chemical LLC S72335MG
Encompass_Preferred
Bromosporine is a broad spectrum inhibitor for bromodomains with IC50 of 0.41 ?M, 0.29 ?M, 0.122 ?M and 0.017 ?M for BRD2, BRD4, BRD9 and CECR2, respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3262