Learn More
Selleck Chemical LLC Bromotetramisole Oxalate
SDP
Catalog No. 5000001524
Encompass_Preferred
(-)-p-Bromotetramisole Oxalate (L-p-Bromotetramisole Oxalate) is a potent and non-specific alkaline phosphatase inhibitor
Catalog No. 50-000-01524 Supplier Selleck Chemical LLC Supplier No. S738310MG
(-)-p-Bromotetramisole Oxalate (L-p-Bromotetramisole Oxalate) is a potent and non-specific alkaline phosphatase inhibitor