Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromothymol Blue, 0.04% (w/v) in Methanol, Ricca Chemical
SDP

Catalog No. R1375500500 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R1375500500 500 mL Natural Poly Bottle
R1375500100 100 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R1375500500 Supplier Ricca Chemical Company Supplier No. R1375500500A
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

pH Indicator

Specifications

Chemical Name or Material Bromothymol Blue, 0.04% (w/v) in Methanol
Melting Point -98°C
Boiling Point ∼64.5°C
CAS 76-59-5, 67-56-1
CAS Min % 99.95, 0.05
CAS Max % 99.95, 0.05
Odor Slight, Alcohol-Like
Packaging Natural Poly Bottle
Molecular Formula CH4O
MDL Number MFCD00004595
Density 0.8
Color Orange/Red
Solubility Information Miscible in water
Quantity 500 mL
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Flash Point 11°C
Physical Form Liquid
SMILES CO
IUPAC Name methanol
Molecular Weight (g/mol) Mixture
Percent Purity 0.04% W/V
Grade Indicator
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.