Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromothymol Blue, 0.04% (w/v) in Methanol, Ricca Chemical
SDP

Catalog No. p-7239418 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R1375500100 100 mL Natural Poly Bottle
R1375500500 500 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R1375500100 Supplier Ricca Chemical Company Supplier No. R1375500100A
Only null left
Add to Cart
Add to Cart

pH Indicator

Specifications

Chemical Name or Material Bromothymol Blue
Name Note 0.04% (w/v) in Methanol
CAS 76-59-5, 67-56-1
Molecular Formula C27H28Br2O5S
Solubility Information Solubility in Water: Infinite
InChI Key NUHCTOLBWMJMLX-UHFFFAOYSA-N
SMILES CC1=C(C(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C(=C4C)Br)O)C(C)C)C(C)C)O)Br
IUPAC Name 2-bromo-4-[3-(3-bromo-4-hydroxy-2-methyl-5-propan-2-ylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-3-methyl-6-propan-2-ylphenol
Molecular Weight (g/mol) Mixture
PubChem CID 6450
ChEBI CHEBI:86155
Percent Purity 0.04% W/V
Grade Indicator
Odor Characteristic alcohol odor
Packaging Natural Poly Bottle
Solvent or Matrix Nonaqueous Solution
Color Orange/Red
Quantity 100 mL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.