Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromothymol Blue, ACS Reagent (Sultone) not water soluble, Reagents

Catalog No. p-200021445
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-336-773 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-336-773 Supplier Reagents Holdings Llc Supplier No. C61070001C2
Only null left
Add to Cart
Add to Cart

Bromothymol Blue, ACS Reagent (Sultone) not water soluble, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Bromothymol Blue, ACS Reagent (Sultone) not water soluble
CAS 76-59-5
Molecular Formula C27H28Br2O5S
Synonym Dibromothymolsulfophthalein, Bromthymol Blue
Molecular Weight (g/mol) 624.3812
Grade ACS
Packaging Glass Bottle
Specific Gravity 1
Color White
Quantity 1 g
Physical Form Solid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.