Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Specifications
Specifications
| Chemical Name or Material | Bromothymol Blue |
| CAS | 34722-90-2, 7732-18-5 |
| Packaging | Natural Poly Bottle |
| Color | Dark Blue |
| pH | 6.70 to 6.90 |
| Quantity | 120 mL |
| Percent Purity | 0.04% W/V |
| Grade | Indicator |
missing translation for 'productTitle'
missing translation for 'byClickingSubmit' missing translation for 'privacyPolicy'.
missing translation for 'spotOppurtunityImprovement'
