Learn More
Bromothymol Blue Indicator, 0.04% w/v, Reagents
Specifications
Specifications
| Chemical Name or Material | Bromothymol Blue Indicator, 0.04% w/v |
| CAS | 7732-18-5, 34722-90-2 |
| Boiling Point | 100°C |
| Packaging | Poly Bottle |
| Specific Gravity | 1.0001 |
| Synonym | Dibromothymolsulfophthalein, Bromthymol Blue |
| Quantity | 100 mL |
| Physical Form | Liquid |
| Percent Purity | 0.04% |
| Grade | Reagent |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.