Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromothymol Blue, Reagent Grade, Innovating Science™
SCI_ED

Catalog No. S000543 Shop All Aldon Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S000543 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S000543 Supplier Aldon Corporation Supplier No. IS11068
Add to Cart
Add to Cart

Chemical Identifiers

CAS 34722-90-2
Molecular Weight (g/mol) 646.39 g/mol

Specifications

Percent Purity 100%
Grade Reagent
Color Violet
Quantity 5 g
Chemical Name or Material Bromothymol Blue, Sodium Salt

This substance or mixture has not been classified as hazardous according to the Globally Harmonized System (GHS) of Classification and Labeling of Chemicals.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.