Learn More
Description
Bromothymol blue dye serves as a pH indicator because of the precise color transition at neutrality. Other applications include: as a vital stain to trace the movement of fluids from the lymph and to define cell walls or nuclei under the microscope for the demonstration of fungal hyphae within plant roots in the assessment of the soluble acidity within single rice grains use in an agar gel medium for enumeration of Bacillus cereus as a component of several bacterial growth and detection substrates such as cystine lactose electrolyte deficient (CLED/BROLACIN); polymyxin pyruvate egg yolk mannitol bromothymol blue (PEMBA) and MacConkey agar.
Specifications
Specifications
| Chemical Name or Material | Bromothymol Blue |
| CAS | 76-59-5 |
| Molecular Weight (g/mol) | 624.38 g/mol |
| Percent Purity | 95% |
| Grade | ACS |
| Color | Purple/Red |
| pH | 6 to 7.6 |
| Quantity | 25 g |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.