Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromothymol Blue, Sigma-Aldrich

Catalog No. 01259036 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-259-036 5 g
01-258-849 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-259-036 Supplier MilliporeSigma Supelco Supplier No. 1144135G
Add to Cart
Add to Cart

Bromothymol blue dye serves as a pH indicator because of the precise color transition at neutrality. Other applications include: as a vital stain to trace the movement of fluids from the lymph and to define cell walls or nuclei under the microscope for the demonstration of fungal hyphae within plant roots in the assessment of the soluble acidity within single rice grains use in an agar gel medium for enumeration of Bacillus cereus as a component of several bacterial growth and detection substrates such as cystine lactose electrolyte deficient (CLED/BROLACIN); polymyxin pyruvate egg yolk mannitol bromothymol blue (PEMBA) and MacConkey agar.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Bromothymol Blue
CAS 76-59-5
Molecular Weight (g/mol) 624.38 g/mol
Percent Purity 95%
Grade ACS
Color Purple/Red
pH 6 to 7.6
Quantity 5 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.