Learn More
Medchemexpress LLC Bromothymol Blue (sodium) | 34722-90-2 | 95.0% | 100 G
SDP
Catalog No. 5000410225
Encompass_Preferred
Bromothymol Blue sodium is a pH indicator used in various biochemical assays. It is particularly useful in pH-shift assays for direct determination of 2-keto-L-gulonic acid (2-KLG) production, exhibiting a clear color change from blue to yellow between pH 6.5-7.5. This chemical compound is provided as a solid with a purity of 95.0% and is intended for research use only.
  • Functions as a pH indicator for biochemical assays
  • Useful in pH-shift assays for detecting 2-keto-L-gulonic acid production
  • Exhibits a clear color change from blue to yellow at pH 6.5-7.5
  • Suitable for kinase assays
  • Presented as a brown to black solid
  • Guaranteed purity of 95.0%
  • Intended for research use only
Catalog No. 50-004-10225 Supplier Medchemexpress LLC Supplier No. HYD0012A100G
Bromothymol Blue sodium is a pH indicator used in various biochemical assays. It is particularly useful in pH-shift assays for direct determination of 2-keto-L-gulonic acid (2-KLG) production, exhibiting a clear color change from blue to yellow between pH 6.5-7.5. This chemical compound is provided as a solid with a purity of 95.0% and is intended for research use only.
  • Functions as a pH indicator for biochemical assays
  • Useful in pH-shift assays for detecting 2-keto-L-gulonic acid production
  • Exhibits a clear color change from blue to yellow at pH 6.5-7.5
  • Suitable for kinase assays
  • Presented as a brown to black solid
  • Guaranteed purity of 95.0%
  • Intended for research use only