Learn More
Medchemexpress LLC Bromothymol Blue sodium | 34722-90-2 | 95.0% | 50 G
SDP
Catalog No. 5000409671
Encompass_Preferred
Bromothymol Blue sodium salt is a pH indicator primarily used for research. It is integral to pH indicator systems, often with potassium phosphate as a buffer, for various characterization and comparison methods. This indicator is particularly effective for direct determination of 2-keto-L-gulonic acid (2-KLG) production due to its distinct color change and optimal pH range of 6.5-7.5, shifting from blue to yellow. It is recommended to store this product protected from light.
  • Functions as a pH indicator in research applications.
  • Enables direct determination of 2-keto-L-gulonic acid (2-KLG) production.
  • Exhibits an obvious color change from blue to yellow in the pH range of 6.5-7.5.
  • Utilized in pH-shift assays.
  • Facilitates visualization of acidification caused by _K. vulgare_ 07 mutants on agar plates.
Catalog No. 50-004-09671 Supplier Medchemexpress LLC Supplier No. HYD0012A50G
Bromothymol Blue sodium salt is a pH indicator primarily used for research. It is integral to pH indicator systems, often with potassium phosphate as a buffer, for various characterization and comparison methods. This indicator is particularly effective for direct determination of 2-keto-L-gulonic acid (2-KLG) production due to its distinct color change and optimal pH range of 6.5-7.5, shifting from blue to yellow. It is recommended to store this product protected from light.
  • Functions as a pH indicator in research applications.
  • Enables direct determination of 2-keto-L-gulonic acid (2-KLG) production.
  • Exhibits an obvious color change from blue to yellow in the pH range of 6.5-7.5.
  • Utilized in pH-shift assays.
  • Facilitates visualization of acidification caused by _K. vulgare_ 07 mutants on agar plates.