Learn More
Bioworld Bromothymol Blue, Sodium Salt, 0.04% (w/v), 4 L
Catalog No. 50255500
Encompass_Preferred
Bromothymol Blue (3',3'-Dibromothymolsulfonphthalein) Sodium Salt, 0.04% (w/v) SolutionA pH indicator which is yellow at pH = 6.0 and blue at pH = 7.6 Appearance: LiquidDensity : 0.99 g/mL at 25 °C(lit.)PH Value: 6.0 - 7.6, yellow to blue
Catalog No. 50-255-500 Supplier Bioworld Supplier No. 402201043
Bromothymol Blue (3',3'-Dibromothymolsulfonphthalein) Sodium Salt, 0.04% (w/v) SolutionA pH indicator which is yellow at pH = 6.0 and blue at pH = 7.6 Appearance: LiquidDensity : 0.99 g/mL at 25 °C(lit.)PH Value: 6.0 - 7.6, yellow to blue