Learn More
Bioworld Bromothymol Blue, Sodium Salt, 0.04% (w/v), 500 mL
Catalog No. 50255498
Encompass_Preferred
Bromothymol Blue (3',3'-Dibromothymolsulfonphthalein) Sodium Salt, 0.04% (w/v) SolutionA pH indicator which is yellow at pH = 6.0 and blue at pH = 7.6 Appearance: LiquidDensity : 0.99 g/mL at 25 °C(lit.)PH Value: 6.0 - 7.6, yellow to blue
Catalog No. 50-255-498 Supplier Bioworld Supplier No. 402201041
Bromothymol Blue (3',3'-Dibromothymolsulfonphthalein) Sodium Salt, 0.04% (w/v) SolutionA pH indicator which is yellow at pH = 6.0 and blue at pH = 7.6 Appearance: LiquidDensity : 0.99 g/mL at 25 °C(lit.)PH Value: 6.0 - 7.6, yellow to blue