Learn More
Bioworld Bromothymol Blue, Sodium Salt, 50 g
Catalog No. 50252557
Encompass_Preferred
Used as a pH indicator. ACS Grade 3,3'-Dibromothymolsulphonephthalein Visual transition interval pH 6.0-7.6
Catalog No. 50-252-557 Supplier Bioworld Supplier No. 217700092
Used as a pH indicator. ACS Grade 3,3'-Dibromothymolsulphonephthalein Visual transition interval pH 6.0-7.6