Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Bromothymol Blue, Sodium Salt
SCI_ED

Catalog No. S25206A Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
5 g
10 g
Packaging:
Reagent Grade
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25206A 10 g Reagent Grade
S25206 5 g Reagent Grade
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S25206A
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CO2/pH indicator

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 34722-90-2
Molecular Formula C27H27Br2NaO5S
Molecular Weight (g/mol) 646.37
MDL Number MFCD00077263,MFCD00077263,MFCD00077263
InChI Key NMKFVGALBGZKGW-FKWCIMQXSA-M
PubChem CID 102183223
IUPAC Name sodium 2-{[(1Z)-3-bromo-2-methyl-4-oxo-5-(propan-2-yl)cyclohexa-2,5-dien-1-ylidene][3-bromo-4-hydroxy-2-methyl-5-(propan-2-yl)phenyl]methyl}benzene-1-sulfonate
SMILES [Na+].CC(C)C1=CC(\C(C2=CC=CC=C2S([O-])(=O)=O)=C2/C=C(C(C)C)C(=O)C(Br)=C2C)=C(C)C(Br)=C1O

Specifications

Assay Percent Range 100% w/v
Linear Formula C27H27Br2O5SNa
Formula Weight 646.04
Grade Reagent
Packaging Reagent Grade
Density approx. 1g/mL
pH Color Changes pH 6.0-7.6 (yellow-blue)
Color Black to Orange
Quantity 10 g
Chemical Name or Material Bromothymol Blue, Sodium Salt
Safety handling Check Alert storage code Red
ShelfLife 5 years
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.