Learn More
Description
This product is for educational use - no C of A is available.
Specifications
Specifications
| Assay Percent Range | 100% w/v |
| Linear Formula | C27H27Br2O5SNa |
| Formula Weight | 646.04 |
| Grade | Reagent |
| Packaging | Reagent Grade |
| Density | approx. 1g/mL |
| pH Color Changes | pH 6.0-7.6 (yellow-blue) |
| Color | Black to Orange |
| Quantity | 5 g |
| Chemical Name or Material | Bromothymol Blue, Sodium Salt |
Safety and Handling
| ShelfLife | 5 years |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
