Learn More
Description
Bromothymol Blue has been used for freshwater pH measurements over a broad range of buffer intensities.
Specifications
Specifications
| Chemical Name or Material | Bromothymol Blue Sodium Salt |
| CAS | 34722-90-2 |
| Color | Blue |
| pH | 6 to 7.6 |
| Quantity | 25 g |
| Molecular Weight (g/mol) | 646.36 g/mol |
| Grade | ACS |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.