Learn More
Bioworld Bromothymol Blue, Sodium Salt, 25 g
Catalog No. 50199197
Encompass_Preferred
Used as a pH indicator. ACS Grade 3,3'-Dibromothymolsulphonephthalein Visual transition interval pH 6.0-7.6
Catalog No. 50-199-197 Supplier Bioworld Supplier No. 217700091
Used as a pH indicator. ACS Grade 3,3'-Dibromothymolsulphonephthalein Visual transition interval pH 6.0-7.6