Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromothymol Blue Solution, BAKER™, J.T. Baker™

Catalog No. 02002473 Shop All Avantor Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
02-002-473 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02-002-473 Supplier Avantor Supplier No. D47207
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A grade of chemicals for laboratory use

Chemical Identifiers

CAS 34722-90-2
Molecular Formula C27H27Br2NaO5S
Molecular Weight (g/mol) 646.37
MDL Number MFCD00077263,MFCD00077263,MFCD00077263
InChI Key NMKFVGALBGZKGW-FKWCIMQXSA-M
PubChem CID 102183223
IUPAC Name sodium 2-{[(1Z)-3-bromo-2-methyl-4-oxo-5-(propan-2-yl)cyclohexa-2,5-dien-1-ylidene][3-bromo-4-hydroxy-2-methyl-5-(propan-2-yl)phenyl]methyl}benzene-1-sulfonate
SMILES [Na+].CC(C)C1=CC(\C(C2=CC=CC=C2S([O-])(=O)=O)=C2/C=C(C(C)C)C(=O)C(Br)=C2C)=C(C)C(Br)=C1O

Specifications

Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 1.00kg
Quantity 500 mL
Chemical Name or Material Bromothymol Blue Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.