Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromothymol Blue Solution R1 0.05% w/v in 20% v/v Reagent Alcohol, Reagents

Catalog No. 01335836
Change view
Click to view available options
Quantity:
120 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-335-836 120 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-335-836 Supplier Reagents Holdings Llc Supplier No. CB004200120A
Add to Cart
Add to Cart

Bromothymol Blue Solution R1 0.05% w/v in 20% v/v Reagent Alcohol, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Bromothymol Blue Solution R1 0.05% w/v in 20% v/v Reagent Alcohol
Boiling Point 98°C
CAS 7732-18-5, 64-17-5, 64-17-5
Synonym Dibromothymolsulfophthalein, Bromthymol Blue
Grade Reagent
Packaging Poly Bottle
Specific Gravity 0.9685
Quantity 120 mL
Physical Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.