Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Bromotrimethylsilane

Catalog No. 92337100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mL
25 mL
100 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
92-337-100ML 100 mL
92-337-25ML 25 mL
92-337-5ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Danger
Catalog No. 92-337-100ML Supplier Sigma Aldrich Supplier No. 92337100ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Bromotrimethylsilane is used with Cl3 to catalyze the direct allylation of a variety of alcohols with allyltrimethylsilane.Bromotrimethylsilane may be used as a derivatizing reagent for the determination of alkyl lysophospholipids in cell culture media using gas chromatography technique.Powerful silylating agent ; Cleavage of ethers ; Smooth cleavage of phosphonic and phosphoric alkyl esters

Specifications

CAS 2857-97-8
Density 1.16 g/mL at 25°C
Boiling Point 79°C
Molecular Formula C3H9BrSi
Refractive Index n20/D 1.424; n20/D 1.424
Linear Formula (CH3)3SiBr
MDL Number MFCD00000048
Quantity 100 mL
Synonym TMBS; Trimethylbromosilane; Trimethylsilyl bromide
Molecular Weight (g/mol) 153.09 g/mol
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.