Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromotrimethylsilane, 98%

Catalog No. AC186650250
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC186650250 25 g Glass bottle
AC186650050 5 g Glass bottle
AC186651000 100 g Glass bottle
AC186655000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. AC186650250 Supplier Thermo Scientific Chemicals Supplier No. 186650250
Only null left
Add to Cart
This item is not returnable. View return policy

CAS: 2857-97-8 | C3H9BrSi | 153.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent, Silylation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2857-97-8
Molecular Formula C3H9BrSi
Molecular Weight (g/mol) 153.1
InChI Key IYYIVELXUANFED-UHFFFAOYSA-N
Synonym trimethylsilyl bromide, trimethylbromosilane, silane, bromotrimethyl, trimethylsilicon bromide, tmsbr, trimethylsilylbromide, bromotrimethyl silane, tmbs, bromo trimethyl silane, tms bromide
PubChem CID 76113
IUPAC Name bromo(trimethyl)silane
SMILES C[Si](C)(C)Br

Specifications

Melting Point -43.0°C
Color Colorless to Yellow
Density 1.1600g/mL
Boiling Point 79.0°C
Flash Point 32°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (Argentometry)
Packaging Glass bottle
Refractive Index 1.4200 to 1.4250
Linear Formula (CH3)3SiBr
Quantity 25 g
Fieser 09,73; 10,59; 12,79; 15,51; 16,50
Specific Gravity 1.16
Solubility Information Solubility in water: decomposes.
Formula Weight 153.1
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Bromotrimethylsilane
Show More Show Less
Hazard Category GHS Signal Word: Danger
EINECSNumber 220-672-

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.