Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Bromotripyrrolidinophosphonium Hexafluorophosphate

Catalog No. 185655G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-565-5G 5 g
18-565-1G 1 g
18-565-25G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Danger
Catalog No. 18-565-5G Supplier Sigma Aldrich Supplier No. 185655G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

It can be used as a coupling reagent:For Suzuki-Miyaura cross-coupling of phenols and arylboronic acids to synthesize biaryls and heterobiaryls.To functionalize pyrimidines (synthesized from 3,4-dihydropyrimidin-2(1H)-ones (DHPMs) via the Kappe dehydrogenation) by using various nucleophiles.To synthesize formamidines by coupling with various primary amines and N,N-diisopropylethylamine.It can also be used as an activating reagent:For the activation of C-OH bond in tautomerizable heterocycles to form the phosphonium salt which aids the Sonogashira coupling of heterocycles with various alkynes.For the one-pot activation of C-O bond in phenols and further coupling reaction with phosphine oxide or phosphite to form C-P bonds.Reactive peptide coupling reagent suitable for +a,+a-dialkyl amino acids; Synthesis of depsipeptidesPyBroP is a registered trademark of Merck KGaA, Darmstadt, Germany

Specifications

CAS 132705-51-2
Molecular Formula C12H24BrF6N3P2
Linear Formula C12H24BrF6N3P2
MDL Number MFCD00077412
Quantity 5 g
Synonym PyBroP(R)
Molecular Weight (g/mol) 466.18 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.