Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Bromoxynil octanoate | 1689-99-2 | 99.3% | 403.11 | 100 MG
SDP

Catalog No. 5000349660
Encompass_Preferred

Bromoxynil octanoate is an herbicide primarily used on maize. It is noted for its potential toxicity to both animals and humans.

  • Herbicide
  • Applicable to maize
  • For research use only
  • Purity: 99.3%
  • Appearance: Solid, white to off-white
  • Soluble in DMSO (100 mg/mL)

Catalog No. 50-003-49660 Supplier Medchemexpress LLC Supplier No. HY136370100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Bromoxynil octanoate is an herbicide primarily used on maize. It is noted for its potential toxicity to both animals and humans.

  • Herbicide
  • Applicable to maize
  • For research use only
  • Purity: 99.3%
  • Appearance: Solid, white to off-white
  • Soluble in DMSO (100 mg/mL)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.