Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Bromperidol | 10457-90-6 | MFCD00866670 | 98.0% | 420.3 g/mol | C21H23BrFNO2 | 10 MG
SDP

Catalog No. 5000298132
Encompass_Preferred

Bromperidol analytical standard provided for research and analytical use. This reference material is intended for method development, calibration, and quality control in laboratory settings. Not for human or clinical use.

  • Analytical standard for research and method validation.
  • High purity: ~98.0%.
  • Molecular formula: C21H23BrFNO2.
  • Molecular weight: 420.3 g/mol.
  • Supplied as a 10 mg reference material; follow SDS for handling and storage.

Catalog No. 50-002-98132 Supplier Medchemexpress LLC Supplier No. HYB0901R10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Bromperidol analytical standard provided for research and analytical use. This reference material is intended for method development, calibration, and quality control in laboratory settings. Not for human or clinical use.

  • Analytical standard for research and method validation.
  • High purity: ~98.0%.
  • Molecular formula: C21H23BrFNO2.
  • Molecular weight: 420.3 g/mol.
  • Supplied as a 10 mg reference material; follow SDS for handling and storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.