Learn More
Medchemexpress LLC Bromperidol hydrochloride | 59453-24-6 | 100.0% | 100 MG
SDP
Catalog No. 5000408960
Encompass_Preferred
Bromperidol hydrochloride (R-11333) is an antipsychotic that shows strong affinity for central dopamine D2 receptors. It also demonstrates synergistic activity against Mycobacteria when combined with Spectinomycin.
  • Exhibits antipsychotic activity
  • High affinity for central dopamine D2 receptors
  • Synergistic action against Mycobacteria when combined with Spectinomycin
  • Antagonizes stereotyped behavior and agitation
  • Inhibits conditioned reactions and intracranial self-stimulation
  • Antagonizes apomorphine-induced emesis and inhibits conditioned avoidance response
Catalog No. 50-004-08960 Supplier Medchemexpress LLC Supplier No. HYB0901A100MG
Bromperidol hydrochloride (R-11333) is an antipsychotic that shows strong affinity for central dopamine D2 receptors. It also demonstrates synergistic activity against Mycobacteria when combined with Spectinomycin.
  • Exhibits antipsychotic activity
  • High affinity for central dopamine D2 receptors
  • Synergistic action against Mycobacteria when combined with Spectinomycin
  • Antagonizes stereotyped behavior and agitation
  • Inhibits conditioned reactions and intracranial self-stimulation
  • Antagonizes apomorphine-induced emesis and inhibits conditioned avoidance response