Learn More
Medchemexpress LLC Bromperidol hydrochloride | 59453-24-6 | 100.0% | 50 MG
SDP
Catalog No. 5000409337
Encompass_Preferred
Bromperidol hydrochloride is an antipsychotic agent that demonstrates high affinity for central dopamine D2 receptors. It can also synergistically kill Mycobacteria when combined with Spectinomycin.
  • High affinity for central dopamine D2 receptors.
  • Antagonizes stereotyped behavior and agitation induced by apomorphine or amphetamine.
  • Inhibits conditioned reactions and learned intracranial self-stimulation in rats.
  • Antagonizes apomorphine-induced emesis.
  • Inhibits the conditioned avoidance response in dogs.
  • Synergistically kills Mycobacteria when combined with Spectinomycin.
Catalog No. 50-004-09337 Supplier Medchemexpress LLC Supplier No. HYB0901A50MG
Bromperidol hydrochloride is an antipsychotic agent that demonstrates high affinity for central dopamine D2 receptors. It can also synergistically kill Mycobacteria when combined with Spectinomycin.
  • High affinity for central dopamine D2 receptors.
  • Antagonizes stereotyped behavior and agitation induced by apomorphine or amphetamine.
  • Inhibits conditioned reactions and learned intracranial self-stimulation in rats.
  • Antagonizes apomorphine-induced emesis.
  • Inhibits the conditioned avoidance response in dogs.
  • Synergistically kills Mycobacteria when combined with Spectinomycin.