Learn More
Medchemexpress LLC Bromperidol hydrochloride | 59453-24-6 | 99.96% | 25 MG
SDP
Catalog No. 5000450461
Encompass_Preferred
Bromperidol hydrochloride is an antipsychotic agent that demonstrates high affinity for central dopamine D2 receptors. It also exhibits synergistic mycobacterial killing activity when combined with Spectinomycin.
  • Exhibits antipsychotic activity.
  • Demonstrates high affinity for central dopamine D2 receptors.
  • Can synergistically kill Mycobacteria when combined with Spectinomycin.
  • Antagonizes stereotyped behavior and agitation induced by apomorphine or amphetamine.
  • Inhibits conditioned reactions and learned intracranial self-stimulation in rats.
  • Antagonizes apomorphine-induced emesis and inhibits conditioned avoidance response in dogs.
Catalog No. 50-004-50461 Supplier Medchemexpress LLC Supplier No. 25MGHYB0901A
Bromperidol hydrochloride is an antipsychotic agent that demonstrates high affinity for central dopamine D2 receptors. It also exhibits synergistic mycobacterial killing activity when combined with Spectinomycin.
  • Exhibits antipsychotic activity.
  • Demonstrates high affinity for central dopamine D2 receptors.
  • Can synergistically kill Mycobacteria when combined with Spectinomycin.
  • Antagonizes stereotyped behavior and agitation induced by apomorphine or amphetamine.
  • Inhibits conditioned reactions and learned intracranial self-stimulation in rats.
  • Antagonizes apomorphine-induced emesis and inhibits conditioned avoidance response in dogs.