Learn More
Medchemexpress LLC Brompheniramine maleate | 980-71-2 | 99.78% | 50 MG
SDP
Catalog No. 502031085
Encompass_Preferred
Brompheniramine maleate is a potent and orally active antihistamine of the alkylamine class. It is a selective histamine H1 receptor antagonist with a Kd of 6.06 nM. It also possesses anticholinergic, antidepressant, and anesthetic properties, and is used for allergic rhinitis research.
  • Blocks hERG channels, calcium channels, and sodium channels
  • Inhibits the amplitude of Ca2+ channel currents in rat ventricular myocytes
  • Blocks muscarinic cholinergic receptors
  • Induces cutaneous analgesia in male Sprague-Dawley rats
Catalog No. 50-203-1085 Supplier Medchemexpress LLC Supplier No. HYB048050MG
Brompheniramine maleate is a potent and orally active antihistamine of the alkylamine class. It is a selective histamine H1 receptor antagonist with a Kd of 6.06 nM. It also possesses anticholinergic, antidepressant, and anesthetic properties, and is used for allergic rhinitis research.
  • Blocks hERG channels, calcium channels, and sodium channels
  • Inhibits the amplitude of Ca2+ channel currents in rat ventricular myocytes
  • Blocks muscarinic cholinergic receptors
  • Induces cutaneous analgesia in male Sprague-Dawley rats