Learn More
Cayman Chemical Brompheniramine (maleate), Purity: ≥95%, CAS: 980-71-2, 50 mg
SDP
Catalog No. 502983861
Encompass_Preferred
A histamine H1 receptor antagonist (Ki = 6.06 nM); also inhibits the reuptake of serotonin and norepinephrine
Catalog No. 50-298-3861 Supplier Cayman Chemical Supplier No. 2099650 MG
A histamine H1 receptor antagonist (Ki = 6.06 nM); also inhibits the reuptake of serotonin and norepinephrine