Learn More
Hach Company Bromphenol Blue Indicator Solution, 100 mL MDB
Supplier:  Hach Company 1455232
Encompass
pH indicator for acidity and alkalinity titrations. Visual transition interval: pH 3.0 (yellow)-pH 4.6 (blue).
Catalog No. 50-199-5071