Learn More
Hach Company Bromphenol Blue Indicator Powder Pillows, pk/100
Supplier: Hach Company 1455099
pH indicator for acidity and alkalinity titrations. Visual transition interval: pH 3.0 (yellow)-pH 4.6 (blue). Sample Size: 50 mL