Learn More
Aqua Solutions Bromothymol Blue 0.04% in Methanol pH 6.0 Yellow - 7.6 Blue (500ML)
SDP
Catalog No. NC0470655
Encompass_Preferred
Bromothymol Blue 0.04% in Methanol pH 6.0 Yellow - 7.6 Blue (500ML)
Catalog No. NC0470655 Supplier Aqua Solutions Supplier No. 1416500ML
Bromothymol Blue 0.04% in Methanol pH 6.0 Yellow - 7.6 Blue (500ML)