Learn More
Hach Company Bromthymol Blue Indicator Solution, 100 mL MDB
Supplier:  Hach Company 25532
Encompass
Bromthymol Blue Indicator Solution. Standardized for colorimetric pH determination. pH range 5.5-8.5. 100 mL Marked Dropping Bottle
Catalog No. 50-199-4218