Learn More
Hach Company Bromthymol Blue Indicator Powder Pillows, pk/100
Supplier:  Hach Company 2067299
Encompass
pH indicator. Visual transition interval: pH 6.0 (yellow)- pH 7.6 (blue).
Catalog No. 50-199-5214