Learn More
Hach Company Bromthymol Blue Indicator Solution, 500 mL
Supplier:  Hach Company 25549
Encompass
Bromthymol Blue Indicator Solution. Standardized for colorimetric pH determination pH range 5.5-8.5. 500 mL bottle
Catalog No. 50-199-4219