Learn More
ATCC Bronchial Epithelial Cell Growth Kit
Catalog No. 502381856
Encompass
Bronchial Epithelial Cell Growth Kit contains components that when added to Airway Epithelial Cell Basal Medium (ATCC PCS-300-030) create a complete ATCC Primary Cell Solutions culture environment for bronchial/tracheal epithelial cells derived from normal human lung (eg, Primary Bronchial/Tracheal Epithelial Cells, Normal, Human, ATCC PCS-300-010). The serum-free medium formulation supports normal bronchial/epithelial cell morphology and promotes rapid growth and proliferation. No feeder layers, extracellular matrix proteins, or other substrates are required.
Catalog No. 50-238-1856 Supplier ATCC Supplier No. PCS300040
Bronchial Epithelial Cell Growth Kit contains components that when added to Airway Epithelial Cell Basal Medium (ATCC PCS-300-030) create a complete ATCC Primary Cell Solutions culture environment for bronchial/tracheal epithelial cells derived from normal human lung (eg, Primary Bronchial/Tracheal Epithelial Cells, Normal, Human, ATCC PCS-300-010). The serum-free medium formulation supports normal bronchial/epithelial cell morphology and promotes rapid growth and proliferation. No feeder layers, extracellular matrix proteins, or other substrates are required.