Learn More
MSC Carlisle Round Trash Can Lid, 20 Gallon, Purple, Polyethylene
Catalog No. 502837521
Encompass
This durable 20 gallon purple round trash can lid is made from extra-thick polyethylene, designed for heavy daily use. It provides a secure seal with snap-down tabs and easy removal via a pull release, effectively containing waste and odors. Resistant to acids, bases, and oils, it tolerates temperatures from -50 to 120 degrees Fahrenheit.
  • Secure seal with snap-down tabs.
  • Easy pull-release removal.
  • Rounded edges and textured surface resist abrasions and scuffs.
  • Raised inner circle for quick stacking.
  • Meets food contact, cleanability, and refuse standards.
  • NSF 2 and NSF 21 certified.
  • Made in the USA.
  • Recyclable HDPE material.
  • Prevents water collection on lid.
  • Aligns with HACCP for color-coded maintenance.
Catalog No. 50-283-7521 Supplier MSC Supplier No. 73919482
This durable 20 gallon purple round trash can lid is made from extra-thick polyethylene, designed for heavy daily use. It provides a secure seal with snap-down tabs and easy removal via a pull release, effectively containing waste and odors. Resistant to acids, bases, and oils, it tolerates temperatures from -50 to 120 degrees Fahrenheit.
  • Secure seal with snap-down tabs.
  • Easy pull-release removal.
  • Rounded edges and textured surface resist abrasions and scuffs.
  • Raised inner circle for quick stacking.
  • Meets food contact, cleanability, and refuse standards.
  • NSF 2 and NSF 21 certified.
  • Made in the USA.
  • Recyclable HDPE material.
  • Prevents water collection on lid.
  • Aligns with HACCP for color-coded maintenance.