Learn More
MSC Carlisle Waste Bin Lid, Polyethylene, 10 Gallon, Round, Black
Catalog No. 502837887
Encompass
The Carlisle Bronco Waste Bin Lid is a durable and secure solution for managing waste. Constructed from extra-thick polyethylene, this flat, round lid is designed to fit 10-gallon containers, providing a hygienic and odor-controlled environment. It features snap-down tab locks for a secure seal and is NSF certified for food safety.
  • Features snap-down tab locks for a secure closure.
  • Constructed from extra-thick polyethylene for durability.
  • NSF certified for food safety and contact.
  • Prevents water from collecting on the lid.
  • Supports HACCP color-coding systems.
  • Resistant to acids, bases, and vegetable oils.
  • Designed for simple removal with a pull release.
  • Textured surface resists abrasions and scuffs.
  • Withstands temperatures from -50 to 120 degrees Fahrenheit.
Catalog No. 50-283-7887 Supplier MSC Supplier No. 73916546
The Carlisle Bronco Waste Bin Lid is a durable and secure solution for managing waste. Constructed from extra-thick polyethylene, this flat, round lid is designed to fit 10-gallon containers, providing a hygienic and odor-controlled environment. It features snap-down tab locks for a secure seal and is NSF certified for food safety.
  • Features snap-down tab locks for a secure closure.
  • Constructed from extra-thick polyethylene for durability.
  • NSF certified for food safety and contact.
  • Prevents water from collecting on the lid.
  • Supports HACCP color-coding systems.
  • Resistant to acids, bases, and vegetable oils.
  • Designed for simple removal with a pull release.
  • Textured surface resists abrasions and scuffs.
  • Withstands temperatures from -50 to 120 degrees Fahrenheit.