Learn More
MSC Carlisle Round Waste Bin Trash Container Lid, 32 Gallon, Red, HDPE
Catalog No. 502838692
Encompass
This round waste bin trash container lid is designed for durability and functionality in various environments. It features a robust construction and food-safe certification, making it suitable for a wide range of waste management applications.
  • Lids snap down securely and remove with ease
  • Durable construction from extra thick plastic for maximum longevity
  • NSF 2 and NSF 21 certified for food safety and color-coding various food products
  • Design prevents water from collecting on the lid
  • Color code program aligns with HACCP practices for sanitary maintenance
  • NSF listed for public health and safety
  • Made in the USA
  • Recyclable, made from high-density polyethylene (HDPE)
Catalog No. 50-283-8692 Supplier MSC Supplier No. 73920076
This round waste bin trash container lid is designed for durability and functionality in various environments. It features a robust construction and food-safe certification, making it suitable for a wide range of waste management applications.
  • Lids snap down securely and remove with ease
  • Durable construction from extra thick plastic for maximum longevity
  • NSF 2 and NSF 21 certified for food safety and color-coding various food products
  • Design prevents water from collecting on the lid
  • Color code program aligns with HACCP practices for sanitary maintenance
  • NSF listed for public health and safety
  • Made in the USA
  • Recyclable, made from high-density polyethylene (HDPE)