Learn More
MSC Carlisle Round Waste Bin Lid, 32 Gallon, HDPE, Flat, Blue
Catalog No. 502838693
Encompass
This Carlisle Bronco Round Waste Bin Lid is designed for 32 gallon trash containers. Made from durable HDPE material, this flat, blue lid is food safe and aligns with HACCP practices, making it suitable for commercial establishments. Its design prevents water from collecting, ensuring hygiene and longevity.
  • Locking lids snap down and press-pull lids stay on and remove with ease.
  • Durable construction made of extra thick plastic for maximum longevity.
  • Certified for storing, separating and color-coding various food products.
  • Design prevents water from collecting on lid.
  • Aligns with HACCP practices and promotes an integrated color-coded sanitary maintenance system.
  • Made in the USA.
  • Recyclable material.
  • Compatible with 32 gallon round trash containers.
Catalog No. 50-283-8693 Supplier MSC Supplier No. 73920241
This Carlisle Bronco Round Waste Bin Lid is designed for 32 gallon trash containers. Made from durable HDPE material, this flat, blue lid is food safe and aligns with HACCP practices, making it suitable for commercial establishments. Its design prevents water from collecting, ensuring hygiene and longevity.
  • Locking lids snap down and press-pull lids stay on and remove with ease.
  • Durable construction made of extra thick plastic for maximum longevity.
  • Certified for storing, separating and color-coding various food products.
  • Design prevents water from collecting on lid.
  • Aligns with HACCP practices and promotes an integrated color-coded sanitary maintenance system.
  • Made in the USA.
  • Recyclable material.
  • Compatible with 32 gallon round trash containers.