Learn More
MSC Carlisle Round Trash Container Lid, 44 gal, HDPE, Flat, Brown
Catalog No. 502838747
Encompass
This round trash container lid is designed for commercial establishments. Made from durable HDPE, this flat lid is suitable for use with 44 gallon trash containers, offering a secure and color-coded solution for waste management or food storage.
  • Flat lid type
  • Round lid shape
  • Compatible with 44 gallon containers
  • Brown color
  • Made from HDPE material
  • 24.64 in diameter
  • 2.24 in height
  • Does not include wheels
  • HACCP, NSF 2, and NSF 21 standards
Catalog No. 50-283-8747 Supplier MSC Supplier No. 73921967
This round trash container lid is designed for commercial establishments. Made from durable HDPE, this flat lid is suitable for use with 44 gallon trash containers, offering a secure and color-coded solution for waste management or food storage.
  • Flat lid type
  • Round lid shape
  • Compatible with 44 gallon containers
  • Brown color
  • Made from HDPE material
  • 24.64 in diameter
  • 2.24 in height
  • Does not include wheels
  • HACCP, NSF 2, and NSF 21 standards