Learn More
MSC Carlisle Bronco Trash Can Lid, 44 Gallon, Flat, White, Polyethylene
Supplier:  MSC 73921975
Encompass
This lid is designed to cover a waste container, keeping contents covered and containing unpleasant odors. It features extra-thick polyethylene construction for durability, a secure snap-down lock, and easy pull release. Its textured surface resists scuffs, and a raised inner circle allows for quick stacking with waste container bases. It withstands temperatures from -50 to 120 degrees Fahrenheit and meets food contact standards.
  • Extra-thick polyethylene construction for daily use.
  • Snap-down tabs for a secure seal.
  • Pull release for simple removal.
  • Textured surface resists abrasions and scuffs.
  • Raised inner circle for quick stacking with waste container bases.
  • Withstands temperatures from -50 to 120 degrees Fahrenheit.
  • Meets food contact, cleanability, and thermoplastic refuse standards.
  • Made in America.
  • NSF Certified.
Catalog No. 50-283-8748